3-(Dimethylamino)-1-[3-(trifluoromethoxy)phenyl]prop-2-en-1-one

Lieferant: Apollo Scientific
Achtung

PC300780-1G
APOSPC300780-1GEA 372 EUR
APOSPC300780-1G
3-(Dimethylamino)-1-[3-(trifluoromethoxy)phenyl]prop-2-en-1-one
3-(Dimethylamino)-1-[3-(trifluoromethoxy)phenyl]prop-2-en-1-one
Formel: C₁₂H₁₂F₃NO₂
Molecular Weight: 259,224 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26939178
CAS-Nummer: 154258-46-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR