Methyl 4-(2,6-dimethylmorpholin-4-yl)-2,3,5-trifluorobenzoate

Lieferant: Apollo Scientific

PC300782-1G PC300782-5G PC300782-250MG
APOSPC300782-1GEA 232 EUR
APOSPC300782-1G APOSPC300782-5G APOSPC300782-250MG
Methyl 4-(2,6-dimethylmorpholin-4-yl)-2,3,5-trifluorobenzoate
Methyl 4-(2,6-dimethylmorpholin-4-yl)-2,3,5-trifluorobenzoate
Formel: C₁₄H₁₆F₃NO₃
Molecular Weight: 303.277 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28103344
CAS-Nummer: 1858251-69-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR