Methyl 2,5-difluoro-4-(4-methylpiperazin-1-yl)benzoate
Lieferant: Apollo Scientific
PC300788-1G
PC300788-5G
PC300788-250MG
APOSPC300788-1GEA
290
EUR
APOSPC300788-1G
APOSPC300788-5G
APOSPC300788-250MG
Methyl 2,5-difluoro-4-(4-methylpiperazin-1-yl)benzoate
Methyl 2,5-difluoro-4-(4-methylpiperazin-1-yl)benzoate
Formel:
C₁₃H₁₆F₂N₂O₂ Molecular Weight: 270.275 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28103349 CAS-Nummer: 1858251-84-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...