2,5-Difluoro-4-(4-methylpiperazin-1-yl)benzoic acid

Lieferant: Apollo Scientific
Achtung

PC300789-1G
APOSPC300789-1GEA 361 EUR
APOSPC300789-1G
2,5-Difluoro-4-(4-methylpiperazin-1-yl)benzoic acid
2,5-Difluoro-4-(4-methylpiperazin-1-yl)benzoic acid
Formel: C₁₂H₁₄F₂N₂O₂
Molecular Weight: 256,249 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28103350
CAS-Nummer: 1430223-87-2
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR