6-(Trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid

Lieferant: Apollo Scientific
Achtung

PC300793-1G PC300793-250MG
APOSPC300793-1GEA 0 EUR
APOSPC300793-1G APOSPC300793-250MG
6-(Trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid
6-(Trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid
Formel: C₉H₅F₃N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09258865
CAS-Nummer: 1018828-69-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR