4-Bromo-2-(2,2-diethoxyethoxy)-1-fluorobenzene

Lieferant: Apollo Scientific

PC300794-1G
APOSPC300794-1GEA 296 EUR
APOSPC300794-1G
4-Bromo-2-(2,2-diethoxyethoxy)-1-fluorobenzene
4-Bromo-2-(2,2-diethoxyethoxy)-1-fluorobenzene
Formel: C₁₂H₁₆BrFO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD26939171
CAS-Nummer: 1777895-41-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR