4-Bromo-1-(2,2-diethoxyethoxy)-2-fluorobenzene

Lieferant: Apollo Scientific

PC300795-1G
APOSPC300795-1GEA 248 EUR
APOSPC300795-1G
4-Bromo-1-(2,2-diethoxyethoxy)-2-fluorobenzene
4-Bromo-1-(2,2-diethoxyethoxy)-2-fluorobenzene
Formel: C₁₂H₁₆BrFO₃
Molecular Weight: 307,156 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19168379
CAS-Nummer: 1271673-78-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR