4-Iodo-3-(trifluoromethyl)-1-H-pyrazole
Lieferant: Apollo Scientific
|
Achtung
|
PC300801-1G
PC300801-5G
PC300801-10G
PC300801-25G
APOSPC300801-1GEA
0
EUR
APOSPC300801-1G
APOSPC300801-5G
APOSPC300801-10G
APOSPC300801-25G
4-Iodo-3-(trifluoromethyl)-1-H-pyrazole
4-Iodo-3-(trifluoromethyl)-1-H-pyrazole
Formel:
C₄H₂F₃IN₂ Schmelzpunkt: 137…141 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08064883 CAS-Nummer: 866638-72-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...