3-Amino-3-[4-(trifluoromethoxy)phenyl]propanoic acid

Lieferant: Apollo Scientific
Achtung

PC300802-1G PC300802-5G
APOSPC300802-1GEA 0 EUR
APOSPC300802-1G APOSPC300802-5G
3-Amino-3-[4-(trifluoromethoxy)phenyl]propanoic acid
3-Amino-3-[4-(trifluoromethoxy)phenyl]propanoic acid
Formel: C₁₀H₁₀F₃NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00764454
CAS-Nummer: 213192-56-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR