2,3,5-Trifluoro-4-methoxybenzonitrile

Lieferant: Apollo Scientific
Achtung

PC300803-100MG
APOSPC300803-100MGEA 325 EUR
APOSPC300803-100MG
2,3,5-Trifluoro-4-methoxybenzonitrile
2,3,5-Trifluoro-4-methoxybenzonitrile
Formel: C₈H₄F₃NO
Molecular Weight: 187,119 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07782079
CAS-Nummer: 916791-82-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR