2,3,5-Trifluoro-4-(4-methylpiperazin-1-yl)benzoic acid

Lieferant: Apollo Scientific
Achtung

PC300806-250MG
APOSPC300806-250MGEA 174 EUR
APOSPC300806-250MG
2,3,5-Trifluoro-4-(4-methylpiperazin-1-yl)benzoic acid
2,3,5-Trifluoro-4-(4-methylpiperazin-1-yl)benzoic acid
Formel: C₁₂H₁₃F₃N₂O₂
Molecular Weight: 274,239 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28103355
CAS-Nummer: 812643-49-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR