N-[4-Bromo-2-nitro-5-(trifluoromethyl)phenyl]acetamide

Lieferant: Apollo Scientific

PC300811-1G
APOSPC300811-1GEA 0 EUR
APOSPC300811-1G
N-[4-Bromo-2-nitro-5-(trifluoromethyl)phenyl]acetamide
N-[4-Bromo-2-nitro-5-(trifluoromethyl)phenyl]acetamide
Formel: C₉H₆BrF₃N₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD09263995
CAS-Nummer: 157554-76-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR