2-Chloro-N-[3-(difluoromethoxy)phenyl]acetamide

Lieferant: Apollo Scientific

PC300820-1G
APOSPC300820-1GEA 183 EUR
APOSPC300820-1G
2-Chloro-N-[3-(difluoromethoxy)phenyl]acetamide
2-Chloro-N-[3-(difluoromethoxy)phenyl]acetamide
Formel: C₉H₈ClF₂NO₂
Molecular Weight: 235,615 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01124483
CAS-Nummer: 302936-08-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR