N-(4-Chlorophenyl)-N'-[4-(trifluoromethoxy)phenyl]urea

Lieferant: Apollo Scientific

PC300822-1G PC300822-5G
APOSPC300822-1GEA 125 EUR
APOSPC300822-1G APOSPC300822-5G
N-(4-Chlorophenyl)-N'-[4-(trifluoromethoxy)phenyl]urea
N-(4-Chlorophenyl)-N'-[4-(trifluoromethoxy)phenyl]urea
Formel: C₁₄H₁₀ClF₃N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD02632654
CAS-Nummer: 54730-71-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR