N-(4-Chlorophenyl)-N'-[3-(difluoromethoxy)phenyl]urea

Lieferant: Apollo Scientific

PC300823-1G
APOSPC300823-1GEA 224 EUR
APOSPC300823-1G
N-(4-Chlorophenyl)-N'-[3-(difluoromethoxy)phenyl]urea
N-(4-Chlorophenyl)-N'-[3-(difluoromethoxy)phenyl]urea
Formel: C₁₄H₁₁ClF₂N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD28148243
CAS-Nummer: 540771-25-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR