4-[3-(Difluoromethoxy)phenyl]-2,4-dihydro-3H-1,2,4-triazol-3-one

Lieferant: Apollo Scientific

PC300826-1G
APOSPC300826-1GEA 373 EUR
APOSPC300826-1G
4-[3-(Difluoromethoxy)phenyl]-2,4-dihydro-3H-1,2,4-triazol-3-one
4-[3-(Difluoromethoxy)phenyl]-2,4-dihydro-3H-1,2,4-triazol-3-one
Formel: C₉H₇F₂N₃O₂
Molecular Weight: 227.168 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28148247
CAS-Nummer: 1858255-65-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR