2-Methyl-3-(1,3,4-oxadiazol-2-yl)-6-(trifluoromethyl)pyridine

Lieferant: Apollo Scientific
Achtung

PC300827-1G
APOSPC300827-1GEA 310 EUR
APOSPC300827-1G
2-Methyl-3-(1,3,4-oxadiazol-2-yl)-6-(trifluoromethyl)pyridine
2-Methyl-3-(1,3,4-oxadiazol-2-yl)-6-(trifluoromethyl)pyridine
Formel: C₉H₆F₃N₃O
Molecular Weight: 229,159 g/mol
Schmelzpunkt: 75…77 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD28148241
CAS-Nummer: 1823496-56-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR