6-Phenyl-4-(trifluoromethyl)thieno[2,3-b]pyridin-3-amine

Lieferant: Apollo Scientific

PC300828-1G
APOSPC300828-1GEA 310 EUR
APOSPC300828-1G
6-Phenyl-4-(trifluoromethyl)thieno[2,3-b]pyridin-3-amine
6-Phenyl-4-(trifluoromethyl)thieno[2,3-b]pyridin-3-amine
Formel: C₁₄H₉F₃N₂S
Molecular Weight: 294,295 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28148251
CAS-Nummer: 104960-57-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR