5-Bromo-2-(1H-pyrrol-1-yl)-3-(trifluoromethyl)pyridine

Lieferant: Apollo Scientific

PC300831-1G
APOSPC300831-1GEA 361 EUR
APOSPC300831-1G
5-Bromo-2-(1H-pyrrol-1-yl)-3-(trifluoromethyl)pyridine
5-Bromo-2-(1H-pyrrol-1-yl)-3-(trifluoromethyl)pyridine
Formel: C₁₀H₆BrF₃N₂
Molecular Weight: 291,067 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28347987
CAS-Nummer: 1858241-88-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR