1-(4-Chlorophenoxy)-4-nitro-2-(trifluoromethyl)benzene

Lieferant: Apollo Scientific

PC300832-1G
APOSPC300832-1GEA 147 EUR
APOSPC300832-1G
1-(4-Chlorophenoxy)-4-nitro-2-(trifluoromethyl)benzene
1-(4-Chlorophenoxy)-4-nitro-2-(trifluoromethyl)benzene
Formel: C₁₃H₇ClF₃NO₃
Molecular Weight: 317,648 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01579545
CAS-Nummer: 21802-08-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR