4-Nitro-1-phenoxy-2-(trifluoromethyl)benzene

Lieferant: Apollo Scientific

PC300833-1G
APOSPC300833-1GEA 125 EUR
APOSPC300833-1G
4-Nitro-1-phenoxy-2-(trifluoromethyl)benzene
4-Nitro-1-phenoxy-2-(trifluoromethyl)benzene
Formel: C₁₃H₈F₃NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD28347989
CAS-Nummer: 6969-95-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR