4-[4-Nitro-2-(trifluoromethyl)phenoxy]benzenesulphonyl chloride

Lieferant: Apollo Scientific

PC300834-1G
APOSPC300834-1GEA 248 EUR
APOSPC300834-1G
4-[4-Nitro-2-(trifluoromethyl)phenoxy]benzenesulphonyl chloride
4-[4-Nitro-2-(trifluoromethyl)phenoxy]benzenesulphonyl chloride
Formel: C₁₃H₇ClF₃NO₅S
Molecular Weight: 381,712 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28347990
CAS-Nummer: 1858255-92-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR