4-[4-Amino-2-(trifluoromethyl)phenoxy]-N-methylbenzenesulphonamide

Lieferant: Apollo Scientific

PC300836-1G
APOSPC300836-1GEA 361 EUR
APOSPC300836-1G
4-[4-Amino-2-(trifluoromethyl)phenoxy]-N-methylbenzenesulphonamide
4-[4-Amino-2-(trifluoromethyl)phenoxy]-N-methylbenzenesulphonamide
Formel: C₁₄H₁₃F₃N₂O₃S
Molecular Weight: 346,325 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28347992
CAS-Nummer: 1858251-99-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR