N,N-Dimethyl-4-[4-nitro-2-(trifluoromethyl)phenoxy]benzenesulphonamide

Lieferant: Apollo Scientific

PC300837-1G
APOSPC300837-1GEA 0 EUR
APOSPC300837-1G
N,N-Dimethyl-4-[4-nitro-2-(trifluoromethyl)phenoxy]benzenesulphonamide
Formel: C₁₅H₁₃F₃N₂O₅S
Lagertemperatur: Raumtemperatur
MDL: MFCD28347993
CAS-Nummer: 1858250-85-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR