4-[4-Amino-2-(trifluoromethyl)phenoxy]-N,N-dimethylbenzenesulphonamide
Lieferant: Apollo Scientific
PC300838-1G
APOSPC300838-1GEA
296
EUR
APOSPC300838-1G
4-[4-Amino-2-(trifluoromethyl)phenoxy]-N,N-dimethylbenzenesulphonamide
4-[4-Amino-2-(trifluoromethyl)phenoxy]-N,N-dimethylbenzenesulphonamide
Formel:
C₁₅H₁₅F₃N₂O₃S Molecular Weight: 360,351 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28347997 CAS-Nummer: 1858240-20-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...