4-[4-Nitro-2-(trifluoromethyl)phenoxy]benzenesulphonamide

Lieferant: Apollo Scientific

PC300839-1G
APOSPC300839-1GEA 296 EUR
APOSPC300839-1G
4-[4-Nitro-2-(trifluoromethyl)phenoxy]benzenesulphonamide
4-[4-Nitro-2-(trifluoromethyl)phenoxy]benzenesulphonamide
Formel: C₁₃H₉F₃N₂O₅S
Molecular Weight: 362,281 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28347994
CAS-Nummer: 1858251-46-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR