4-[4-Amino-2-(trifluoromethyl)phenoxy]benzenesulphonamide

Lieferant: Apollo Scientific

PC300840-1G
APOSPC300840-1GEA 296 EUR
APOSPC300840-1G
4-[4-Amino-2-(trifluoromethyl)phenoxy]benzenesulphonamide
4-[4-Amino-2-(trifluoromethyl)phenoxy]benzenesulphonamide
Formel: C₁₃H₁₁F₃N₂O₃S
Molecular Weight: 332,298 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28347996
CAS-Nummer: 1858252-06-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR