4-{[5-(Trifluoromethyl)pyridin-2-yl]oxy}benzenesulphonamide

Lieferant: Apollo Scientific

PC300842-1G
APOSPC300842-1GEA 310 EUR
APOSPC300842-1G
4-{[5-(Trifluoromethyl)pyridin-2-yl]oxy}benzenesulphonamide
4-{[5-(Trifluoromethyl)pyridin-2-yl]oxy}benzenesulphonamide
Formel: C₁₂H₉F₃N₂O₃S
Molecular Weight: 318,272 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07774163
CAS-Nummer: 874839-04-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR