N-Methyl-4-{[4-(trifluoromethyl)pyrimidin-2-yl]oxy}benzenesulphonamide

Lieferant: Apollo Scientific

PC300844-1G
APOSPC300844-1GEA 361 EUR
APOSPC300844-1G
N-Methyl-4-{[4-(trifluoromethyl)pyrimidin-2-yl]oxy}benzenesulphonamide
N-Methyl-4-{[4-(trifluoromethyl)pyrimidin-2-yl]oxy}benzenesulphonamide
Formel: C₁₂H₁₀F₃N₃O₃S
Molecular Weight: 333,286 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11109534
CAS-Nummer: 1858256-57-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR