4-{[4-(Trifluoromethyl)pyrimidin-2-yl]oxy}benzenesulphonamide
Lieferant: Apollo Scientific
PC300845-1G
APOSPC300845-1GEA
361
EUR
APOSPC300845-1G
4-{[4-(Trifluoromethyl)pyrimidin-2-yl]oxy}benzenesulphonamide
4-{[4-(Trifluoromethyl)pyrimidin-2-yl]oxy}benzenesulphonamide
Formel:
C₁₁H₈F₃N₃O₃S Molecular Weight: 319,26 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11109535 CAS-Nummer: 1858257-31-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...