2-[2-(6-Fluoroimidazo[1,2-a]pyridin-2-yl)ethyl]-1H-isoindole-1,3(2H)-dione

Lieferant: Apollo Scientific

PC300848-1G
APOSPC300848-1GEA 371 EUR
APOSPC300848-1G
2-[2-(6-Fluoroimidazo[1,2-a]pyridin-2-yl)ethyl]-1H-isoindole-1,3(2H)-dione
2-[2-(6-Fluoroimidazo[1,2-a]pyridin-2-yl)ethyl]-1H-isoindole-1,3(2H)-dione
Formel: C₁₇H₁₂FN₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD27960210
CAS-Nummer: 1417636-77-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR