3-(4-Fluorophenyl)-3-(1-oxo-1,3-dihydro-2H-isoindol-2-yl)propanoic acid

Lieferant: Apollo Scientific

PC300849-1G
APOSPC300849-1GEA 241 EUR
APOSPC300849-1G
3-(4-Fluorophenyl)-3-(1-oxo-1,3-dihydro-2H-isoindol-2-yl)propanoic acid
3-(4-Fluorophenyl)-3-(1-oxo-1,3-dihydro-2H-isoindol-2-yl)propanoic acid
Formel: C₁₇H₁₄FNO₃
Molecular Weight: 299,296 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02571416
CAS-Nummer: 478260-10-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR