N-(3-Chlorophenyl)-N'-[2-methyl-6-(trifluoromethyl)pyridin-3-yl]urea

Lieferant: Apollo Scientific

PC300851-1G
APOSPC300851-1GEA 433 EUR
APOSPC300851-1G
N-(3-Chlorophenyl)-N'-[2-methyl-6-(trifluoromethyl)pyridin-3-yl]urea
N-(3-Chlorophenyl)-N'-[2-methyl-6-(trifluoromethyl)pyridin-3-yl]urea
Formel: C₁₄H₁₁ClF₃N₃O
Molecular Weight: 329,705 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28954643
CAS-Nummer: 1858256-58-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR