4-[4-(Piperidin-1-ylsulphonyl)phenoxy]-3-(trifluoromethyl)aniline

Lieferant: Apollo Scientific

PC300854-1G
APOSPC300854-1GEA 371 EUR
APOSPC300854-1G
4-[4-(Piperidin-1-ylsulphonyl)phenoxy]-3-(trifluoromethyl)aniline
4-[4-(Piperidin-1-ylsulphonyl)phenoxy]-3-(trifluoromethyl)aniline
Formel: C₁₈H₁₉F₃N₂O₃S
Molecular Weight: 400,415 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28954646
CAS-Nummer: 1858252-16-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR