N-[2-Methyl-6-(trifluoromethyl)pyridin-3-yl]-N'-(tetrahydrofuran-2-ylmethyl)urea

Lieferant: Apollo Scientific

PC300856-1G
APOSPC300856-1GEA 420 EUR
APOSPC300856-1G
N-[2-Methyl-6-(trifluoromethyl)pyridin-3-yl]-N'-(tetrahydrofuran-2-ylmethyl)urea
N-[2-Methyl-6-(trifluoromethyl)pyridin-3-yl]-N'-(tetrahydrofuran-2-ylmethyl)urea
Formel: C₁₃H₁₆F₃N₃O₂
Molecular Weight: 303,28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28954652
CAS-Nummer: 1858240-24-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR