1-[2,6-Dichloro-4-(trifluoromethyl)phenyl]-N-methyl-1H-pyrrole-2-sulphonamide

Lieferant: Apollo Scientific

PC300857-1G
APOSPC300857-1GEA 371 EUR
APOSPC300857-1G
1-[2,6-Dichloro-4-(trifluoromethyl)phenyl]-N-methyl-1H-pyrrole-2-sulphonamide
1-[2,6-Dichloro-4-(trifluoromethyl)phenyl]-N-methyl-1H-pyrrole-2-sulphonamide
Formel: C₁₂H₉Cl₂F₃N₂O₂S
Molecular Weight: 373,178 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28954653
CAS-Nummer: 1858251-13-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR