N-[4-Phenoxy-2-(trifluoromethyl)phenyl]acetamide

Lieferant: Apollo Scientific

PC300858-1G
APOSPC300858-1GEA 371 EUR
APOSPC300858-1G
N-[4-Phenoxy-2-(trifluoromethyl)phenyl]acetamide
N-[4-Phenoxy-2-(trifluoromethyl)phenyl]acetamide
Formel: C₁₅H₁₂F₃NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD28954654
CAS-Nummer: 1858255-86-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR