4-[6-(Trifluoromethyl)imidazo[1,2-a]pyridin-2-yl]benzonitrile

Lieferant: Apollo Scientific

PC300864-1G
APOSPC300864-1GEA 451 EUR
APOSPC300864-1G
4-[6-(Trifluoromethyl)imidazo[1,2-a]pyridin-2-yl]benzonitrile
4-[6-(Trifluoromethyl)imidazo[1,2-a]pyridin-2-yl]benzonitrile
Formel: C₁₅H₈F₃N₃
Molecular Weight: 287,239 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28992256
CAS-Nummer: 907960-23-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR