N-[2-Methyl-6-(trifluoromethyl)pyridin-3-yl]-N'-(3-pyrrolidin-1-ylphenyl)urea

Lieferant: Apollo Scientific

PC300868-1G
APOSPC300868-1GEA 493 EUR
APOSPC300868-1G
N-[2-Methyl-6-(trifluoromethyl)pyridin-3-yl]-N'-(3-pyrrolidin-1-ylphenyl)urea
N-[2-Methyl-6-(trifluoromethyl)pyridin-3-yl]-N'-(3-pyrrolidin-1-ylphenyl)urea
Formel: C₁₈H₁₉F₃N₄O
Lagertemperatur: Raumtemperatur
MDL: MFCD28992246
CAS-Nummer: 1980064-08-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR