N-(3-Fluoro-4-methoxybenzyl)-N'-[2-methyl-6-(trifluoromethyl)pyridin-3-yl]urea

Lieferant: Apollo Scientific

PC300869-1G
APOSPC300869-1GEA 618 EUR
APOSPC300869-1G
N-(3-Fluoro-4-methoxybenzyl)-N'-[2-methyl-6-(trifluoromethyl)pyridin-3-yl]urea
N-(3-Fluoro-4-methoxybenzyl)-N'-[2-methyl-6-(trifluoromethyl)pyridin-3-yl]urea
Formel: C₁₆H₁₅F₄N₃O₂
Molecular Weight: 357,303 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28992247
CAS-Nummer: 1980064-17-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR