1,1,1-Trifluoro-3-[(tetrahydrofuran-2-ylmethyl)amino]propan-2-ol

Lieferant: Apollo Scientific
Achtung

PC300872-1G
APOSPC300872-1GEA 400 EUR
APOSPC300872-1G
1,1,1-Trifluoro-3-[(tetrahydrofuran-2-ylmethyl)amino]propan-2-ol
1,1,1-Trifluoro-3-[(tetrahydrofuran-2-ylmethyl)amino]propan-2-ol
Formel: C₈H₁₄F₃NO₂
Molecular Weight: 213,197 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05256299
CAS-Nummer: 866135-63-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR