3-[(4-Chlorobenzyl)amino]-1,1,1-trifluoropropan-2-ol
Lieferant: Apollo Scientific
PC300876-1G
APOSPC300876-1GEA
400
EUR
APOSPC300876-1G
3-[(4-Chlorobenzyl)amino]-1,1,1-trifluoropropan-2-ol
3-[(4-Chlorobenzyl)amino]-1,1,1-trifluoropropan-2-ol
Formel:
C₁₀H₁₁ClF₃NO Molecular Weight: 253,649 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01871713 CAS-Nummer: 478081-30-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...