1,1,1-Trifluoro-3-morpholin-4-ylpropan-2-ol

Lieferant: Apollo Scientific
Achtung

PC300877-1G
APOSPC300877-1GEA 163 EUR
APOSPC300877-1G
1,1,1-Trifluoro-3-morpholin-4-ylpropan-2-ol
1,1,1-Trifluoro-3-morpholin-4-ylpropan-2-ol
Formel: C₇H₁₂F₃NO₂
Molecular Weight: 199,171 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00551088
CAS-Nummer: 478068-15-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR