N,N-Diisopropyl-4-(trifluoromethoxy)benzenesulphonamide

Lieferant: Apollo Scientific

PC300883-10G PC300883-1G PC300883-5G
APOSPC300883-10GEA 868 EUR
APOSPC300883-10G APOSPC300883-1G APOSPC300883-5G
N,N-Diisopropyl-4-(trifluoromethoxy)benzenesulphonamide
N,N-Diisopropyl-4-(trifluoromethoxy)benzenesulphonamide
Formel: C₁₃H₁₈F₃NO₃S
Molecular Weight: 325.347 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28992264
CAS-Nummer: 1993322-94-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR