N-(3-Fluoro-2-methylphenyl)acetamide

Lieferant: Apollo Scientific

PC300885-1G
APOSPC300885-1GEA 114 EUR
APOSPC300885-1G
N-(3-Fluoro-2-methylphenyl)acetamide
N-(3-Fluoro-2-methylphenyl)acetamide
Formel: C₉H₁₀FNO
Lagertemperatur: Raumtemperatur
MDL: MFCD03787362
CAS-Nummer: 322-33-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR