N-(3-Fluoro-2-methyl-6-nitrophenyl)acetamide

Lieferant: Apollo Scientific

PC300886-1G
APOSPC300886-1GEA 171 EUR
APOSPC300886-1G
N-(3-Fluoro-2-methyl-6-nitrophenyl)acetamide
N-(3-Fluoro-2-methyl-6-nitrophenyl)acetamide
Formel: C₉H₉FN₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD03787363
CAS-Nummer: 156243-41-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR