1-[3-(Difluoromethoxy)phenyl]-1H-pyrrole-2,5-dione

Lieferant: Apollo Scientific

PC300887-1G
APOSPC300887-1GEA 435 EUR
APOSPC300887-1G
1-[3-(Difluoromethoxy)phenyl]-1H-pyrrole-2,5-dione
1-[3-(Difluoromethoxy)phenyl]-1H-pyrrole-2,5-dione
Formel: C₁₁H₇F₂NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD29918590
CAS-Nummer: 82673-81-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR