2-(3-Chloro-4-fluorophenyl)imidazo[1,2-a]pyridine-7-carboxylic acid

Lieferant: Apollo Scientific

PC300895-1G
APOSPC300895-1GEA 621 EUR
APOSPC300895-1G
2-(3-Chloro-4-fluorophenyl)imidazo[1,2-a]pyridine-7-carboxylic acid
2-(3-Chloro-4-fluorophenyl)imidazo[1,2-a]pyridine-7-carboxylic acid
Formel: C₁₄H₈ClFN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD29918594
CAS-Nummer: 1493735-34-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR