N-[4-Bromo-2-(trifluoromethoxy)phenyl]azepane-1-carbothioamide

Lieferant: Apollo Scientific

PC300901-1G
APOSPC300901-1GEA 497 EUR
APOSPC300901-1G
N-[4-Bromo-2-(trifluoromethoxy)phenyl]azepane-1-carbothioamide
N-[4-Bromo-2-(trifluoromethoxy)phenyl]azepane-1-carbothioamide
Formel: C₁₄H₁₆BrF₃N₂OS
Lagertemperatur: Raumtemperatur
MDL: MFCD30188170
CAS-Nummer: 2197054-46-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR