N-[4-Bromo-2-(trifluoromethoxy)phenyl]-N'-methylthiourea

Lieferant: Apollo Scientific

PC300903-1G
APOSPC300903-1GEA 497 EUR
APOSPC300903-1G
N-[4-Bromo-2-(trifluoromethoxy)phenyl]-N'-methylthiourea
N-[4-Bromo-2-(trifluoromethoxy)phenyl]-N'-methylthiourea
Formel: C₉H₈BrF₃N₂OS
Lagertemperatur: Raumtemperatur
MDL: MFCD30188166
CAS-Nummer: 1988424-15-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR